Learn More
Enzo Life Sciences LAG-3 (human) : Fc (human), (recombinant) (50 µg)
SDP
Supplier:  Enzo Life Sciences ALX522078C050
Encompass_Preferred
Alternative name: Lymphocyte activation gene 3, FDC protein, CD223. MW: ~80kDa (SDS-PAGE). Endotoxin content: <0.1EU/mg purified protein (LAL test; Bio Whittaker). Purity: ≥99% (SDS-PAGE). Formulation: Liquid. In PBS. Source: Produced in CHO cells. The sequence coding for the 4 extracellular Ig-like domains of human LAG-3 (D1-D4) is fused to the Fc portion of human IgG1. Specificity: Binds to human, mouse and monkey MHC class II. Use/Stability: Stable for at least 6 months after receipt when stored at -20°C. UniProt: P18627. Application: ELISA. Flow Cytometry. Long Term Storage: -20°C
Catalog No. NC0489298