Learn More
ACROBiosystems ACROBiosystems Cynomolgus LAG-3 / CD223 Protein, Mouse IgG2a Fc Tag
Catalog No. 501957227
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus LAG-3, Mouse IgG2a Fc tag (LA3-C52A0) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - His 449 (Accession # XP_005570011.1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 73.5 kDa. The protein migrates as 95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus LAG-3, Mouse IgG2a Fc tag (Cat. No. LA3-C52A0) at 2 μg/mL (100 μL/well) can bind Madarex LAG-3 Mab, Human IgG1 with a linear range of 20-78 ng/mL (QC tested).
Catalog No. 50-195-7227 Supplier ACROBiosystems Supplier No. LA3C52A01MG
Recombinant Protein;1MG;Cynomolgus LAG-3, Mouse IgG2a Fc tag (LA3-C52A0) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - His 449 (Accession # XP_005570011.1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 73.5 kDa. The protein migrates as 95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus LAG-3, Mouse IgG2a Fc tag (Cat. No. LA3-C52A0) at 2 μg/mL (100 μL/well) can bind Madarex LAG-3 Mab, Human IgG1 with a linear range of 20-78 ng/mL (QC tested).