Learn More
Enzo Life Sciences LAG-3 (mouse) : Fc (mouse), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ALX522099C050
Encompass_Preferred
Alternative name: Lymphocyte activation gene 3, FDC protein, CD223. Concentration: 0.5mg/ml. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In PBS. Source: Produced in CHO cells. The sequence coding for the 4 extracellular Ig-like domains of mouse LAG-3 (D1-D4) is fused to the Fc portion of mouse IgG2a (hinge-CH2-CH3). Specificity: Binds to human and mouse MHC class II. Long Term Storage: -20°C. Applications: Flow Cytometry .
Catalog No. 50-200-9417