Learn More
Aviva Systems Biology Corp LAG3 Antibody - C-terminal region (ARP59142_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59142_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LAG3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human LAG3. Alternate names: CD223ApplicationWB. Species reactivity: Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-286-592