Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAG3 Detection Set, ProSci
SDP

Catalog No. 16595325 Shop All ProSci Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-325 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-595-325 Supplier ProSci LLC Supplier No. SD8700SET
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Llama Monoclonal Detection Sets

LAG3 Monoclonal specifically detects LAG3 in Human samples. It is validated for ELISA, IHC-P, IF, Flow

The lymphocyte activation gene-3 (LAG3) is a member of the immunoglobulin superfamily and binds MHC class II with high affinity (1), negatively regulating T-cell function and homeostasis (2). It is expressed in B, T, and NK cells, monocytes, and dendritic cells (3), and acts to regulate T cell expansion (4). LAG3 is also an important immune checkpoint protein, with anti-LAG3 antibodies activating T effector cells and affecting regulatory T cell functions. Furthermore LAG3 appears to act in a synergistic fashion with PD-1/PD-L1, suggesting that a dual antibody approach may prove useful in cancer immunotherapy.

Specifications

Antigen LAG3
Applications ELISA, Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Concentration Antibody 1 mg/mL
Conjugate Unconjugated
Formulation LAG3 Antibodies are supplied in PBS.
Host Species Llama
Purification Method LAG3 Antibodies are affinity chromatography purified via Nickel column. Antibodies are supplied as a His-tagged purified protein. They also contains a myc-tag for detection.
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage LAG3 antibodies should be stored in working aliquots at -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Product Type Detection Sets
Form Liquid
Isotype Not Applicable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.