Learn More
Sino Biological Mouse LAG3 / CD223 / Lymphocyte activation gene 3 Protein (ECD) 50ug
Supplier:  Sino Biological 53069MNAH50
Encompass_Preferred
A DNA sequence encoding the mouse LAG3 (NP_032505.1) (Met1-Leu442) was expressed.
Catalog No. 50-211-5580