Learn More
ACROBiosystems Recombinant Protein;Human LAG3 / CD223 Protein, Fc Tag (HPLC-verified);HEK293;100UG;LA3,LAG3,CD223,LAG-3,FDC
Supplier:  ACROBiosystems LA3H5255100UG
Encompass_Preferred
Recombinant Protein;Human LAG3 / CD223 Protein, Fc Tag (HPLC-verified);100UG;Human LAG-3, Fc Tag (HPLC-verified) (LA3-H5255) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accession # P18627-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 72.8 kDa. The protein migrates as 80-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LAG-3, Fc Tag (HPLC-verified) (Cat. No. LA3-H5255) at 2 μg/mL (100 μL/well) can bind Madarex LAG-3 MAb with a linear range of 0.4-3 ng/mL (QC tested).
Catalog No. 50-165-7495