Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human LAG-3 / CD223 Protein, Fc Tag, Avi Tag (Avitag™);HEK293;200UG;LA3,LAG3,CD223,LAG-3,FDC
Supplier:  ACROBiosystems LA3H82FB200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human LAG-3 / CD223 Protein, Fc Tag, Avi Tag (Avitag™);200UG;Biotinylated Human LAG-3, Fc Tag, Avi Tag (LA3-H82Fb) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accession # P18627-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 75.0 kDa. The protein migrates as 90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Human LAG3 MAb, Human IgG1 at 2 μg/mL (100 μL/well) can bind Biotinylated Human LAG-3, Fc Tag, Avi Tag (Cat. No. LA3-H82Fb) with a linear range of 0.1-3 ng/mL (QC tested).
Catalog No. 50-165-7459