Learn More
ACROBiosystems Recombinant Protein;Rat LAG3 / CD223 Protein;HEK293;1MG;LA3,LAG3,CD223,LAG-3,FDC
Supplier:  ACROBiosystems LA3R52H51MG
Encompass_Preferred
Recombinant Protein;Rat LAG3 / CD223 Protein;1MG;Rat LAG-3, His Tag (LA3-R52H5) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Leu 442 (Accession # NP_997678).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.6 kDa. The protein migrates as 55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8642