Learn More
ACROBiosystems Recombinant Protein;Human LAG-3 / CD223 Protein, Llama IgG2b Fc Tag, low endotoxin;HEK293;100UG;LA3,LAG3,CD223,LAG-3,FDC
Supplier:  ACROBiosystems LA3H525C100UG
Encompass_Preferred
Recombinant Protein;Human LAG-3 / CD223 Protein, Llama IgG2b Fc Tag, low endotoxin;100UG;Human LAG-3, Llama IgG2b Fc Tag, low endotoxin (LA3-H525c) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accession # P18627-1).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 74.0 kDa. The protein migrates as 90 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LAG-3, Llama IgG2b Fc Tag, low endotoxin (Cat. No. LA3-H525c) at 2 μg/mL (100 μL/well) can bind Anti-Human LAG3 MAb with a linear range of 0.2-4 ng/mL (QC tested).
Catalog No. 50-165-7690