Learn More
ACROBiosystems Human LAG-3 Protein, Mouse IgG2a Fc Tag, Low Endotoxin, 1 mg
Catalog No. 501658376
Encompass_Preferred
This human LAG-3 (CD223) protein is a member of the immunoglobulin superfamily, containing four extracellular Ig-like domains, and is involved in lymphocyte activation through its binding to HLA class-II antigens. The protein is expressed in human HEK293 cells and contains amino acids Leu 23 to Leu 450 with a C-terminal mouse IgG2a Fc tag. It is supplied as a lyophilized powder and features low endotoxin levels.
  • Greater than 95% purity as determined by both SDS-PAGE and SEC-MALS
  • Calculated molecular weight of 73.3 kDa, migrating as 90-100 kDa under reducing conditions due to glycosylation
  • Formulated in Tris with Glycine, Arginine, NaCl, and trehalose at pH 7.5
  • Endotoxin level is less than 0.01 EU per μg
  • Expressed from human 293 cells
Catalog No. 50-165-8376 Supplier ACROBiosystems Supplier No. LA3H52AA1MG
This human LAG-3 (CD223) protein is a member of the immunoglobulin superfamily, containing four extracellular Ig-like domains, and is involved in lymphocyte activation through its binding to HLA class-II antigens. The protein is expressed in human HEK293 cells and contains amino acids Leu 23 to Leu 450 with a C-terminal mouse IgG2a Fc tag. It is supplied as a lyophilized powder and features low endotoxin levels.
  • Greater than 95% purity as determined by both SDS-PAGE and SEC-MALS
  • Calculated molecular weight of 73.3 kDa, migrating as 90-100 kDa under reducing conditions due to glycosylation
  • Formulated in Tris with Glycine, Arginine, NaCl, and trehalose at pH 7.5
  • Endotoxin level is less than 0.01 EU per μg
  • Expressed from human 293 cells