Learn More
Medchemexpress LLC LAGIPRA peptide (TFA) | 98.2% | 5756.50 (free base) | 5 MG
SDP
Catalog No. 5000424692
Encompass_Preferred
LAGIPRA peptide TFA is a long-acting GIP1R agonist that enhances insulin sensitivity by augmenting glucose disposal. It also reduces branched-chain amino acids (BCAAs) and ketoacids, making it a potential subject for research into type 2 diabetes.
  • Long-acting GIP1R agonist
  • Enhances insulin sensitivity by augmenting glucose disposal
  • Reduces branched-chain amino acids (BCAAs) and ketoacids
  • Potential for research into type 2 diabetes
Catalog No. 50-004-24692 Supplier Medchemexpress LLC Supplier No. HYP11285A5MG
LAGIPRA peptide TFA is a long-acting GIP1R agonist that enhances insulin sensitivity by augmenting glucose disposal. It also reduces branched-chain amino acids (BCAAs) and ketoacids, making it a potential subject for research into type 2 diabetes.
  • Long-acting GIP1R agonist
  • Enhances insulin sensitivity by augmenting glucose disposal
  • Reduces branched-chain amino acids (BCAAs) and ketoacids
  • Potential for research into type 2 diabetes