Learn More
Cayman Chemical L-α-Hydroxyglutaric Acid (sodium salt), Purity: ≥95%, CAS: 63512-50-5, 50 mg
SDP
Catalog No. 502951986
Encompass_Preferred
An α-hydroxy acid; metabolized to α-ketoglutarate by L-2-hydroxyglutarate dehydrogenase; levels are increased in 2-hydroxyglutaric aciduria; competitively inhibits α-ketoglutarate-dependent dioxygenases, including several involved in histone lysine and DNA demethylation
Catalog No. 50-295-1986 Supplier Cayman Chemical Supplier No. 1187650 MG
An α-hydroxy acid; metabolized to α-ketoglutarate by L-2-hydroxyglutarate dehydrogenase; levels are increased in 2-hydroxyglutaric aciduria; competitively inhibits α-ketoglutarate-dependent dioxygenases, including several involved in histone lysine and DNA demethylation