Learn More
ACROBiosystems ACROBiosystems Mouse LAIR1 / CD305 Protein, Mouse IgG2a Fc Tag, low endotoxin
Supplier:  ACROBiosystems LA1M52541MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse LAIR-1, Mouse IgG2a Fc Tag, low endotoxin (LA1-M5254) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Tyr 141 (Accession # Q8BG84-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 40.6 kDa. The protein migrates as 50-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Collagen I protein at 2 μg/mL (100 μL/well) can bind Mouse LAIR-1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. LA1-M5254) with a linear range of 1-10 ng/mL (QC tested).
Catalog No. 50-195-7234