Learn More
ACROBiosystems ACROBiosystems Rhesus macaque LAIR-1 / CD305 Protein, Mouse IgG2a Fc Tag
Supplier:  ACROBiosystems LA1C52A31MG
Encompass_Preferred
Recombinant Protein;1MG;Rhesus macaque LAIR-1, Mouse IgG2a Fc Tag (LA1-C52A3) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Try 165 (Accession # ALV66454.1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 42.9 kDa. The protein migrates as 64 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Collagen I protein at 2 μg/mL (100 μL/well) can bind Rhesus macaque LAIR-1, Mouse IgG2a Fc Tag (Cat. No. LA1-C52A3) with a linear range of 5-78 ng/mL (QC tested).
Catalog No. 50-195-7248