Learn More
Medchemexpress LLC LAIR1 Human Biotin 100ug
SDP
Catalog No. 5000181260
Encompass_Preferred
LAIR1 protein has a negative regulatory effect on NK cells B cells and T cells It activates the phosphatases PTPN6 and PTPN11 inhibits calcium increases in B cells and regulates cytokine production in CD4 T cells LAIR1 Protein Human (Biotinylated HEK293 Avi-His) is the recombinant human-derived LAIR1 protein expressed by HEK293 with C-Avi C-6 His labeled tag
Catalog No. 50-001-81260 Supplier Medchemexpress LLC Supplier No. HYP72390100UG
LAIR1 protein has a negative regulatory effect on NK cells B cells and T cells It activates the phosphatases PTPN6 and PTPN11 inhibits calcium increases in B cells and regulates cytokine production in CD4 T cells LAIR1 Protein Human (Biotinylated HEK293 Avi-His) is the recombinant human-derived LAIR1 protein expressed by HEK293 with C-Avi C-6 His labeled tag