Learn More
ACROBiosystems Recombinant Protein;Human LAIR1 / CD305 Protein, Fc Tag;HEK293;1MG;LA1,LAIR1,CD305,LAIR-1
Catalog No. 501658276
Encompass_Preferred
Recombinant Protein;Human LAIR1 / CD305 Protein, Fc Tag;1MG;Human LAIR-1, Fc Tag (LA1-H5252) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - His 163 (Accession # Q6GTX8-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.1 kDa. The protein migrates as 55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8276 Supplier ACROBiosystems Supplier No. LA1H52521MG
Recombinant Protein;Human LAIR1 / CD305 Protein, Fc Tag;1MG;Human LAIR-1, Fc Tag (LA1-H5252) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - His 163 (Accession # Q6GTX8-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.1 kDa. The protein migrates as 55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.