Learn More
ACROBiosystems Recombinant Protein;Human LAIR1 / CD305 Protein, His Tag;HEK293;1MG;CD5,LAIR1,CD305,LAIR-1
Supplier:  ACROBiosystems CD5H52H11MG
Encompass_Preferred
Recombinant Protein;Human LAIR1 / CD305 Protein, His Tag;1MG;Human LAIR-1, His Tag (CD5-H52H1) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - His 163 (Accession # Q6GTX8-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.4 kDa. The protein migrates as 30-36 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8258