Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human LAIR1 / CD305 Protein, His Tag & Avi Tag (Avitag™);HEK293;25UG;LA1,LAIR1,CD305,LAIR-1
Supplier: ACROBiosystems LA1H82E325UG
Recombinant Protein;Biotinylated Human LAIR1 / CD305 Protein, His Tag & Avi Tag (Avitag™);25UG;Biotinylated Human LAIR-1, His Tag, Avi Tag (LA1-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - His 163 (Accession # Q6GTX8-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 19.2 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.