Learn More
ACROBiosystems Recombinant Protein;Rhesus macaque LAIR1 / CD305 Protein, Mouse IgG2a Fc Tag;HEK293;100UG;LA1,LAIR1,CD305,LAIR-1
Catalog No. 501658174
Encompass_Preferred
Recombinant Protein;Rhesus macaque LAIR1 / CD305 Protein, Mouse IgG2a Fc Tag;100UG;Rhesus macaque LAIR-1, mouse IgG2a Fc tag (LA1-C52A3) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Try 165 (Accession # ALV66454.1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 42.9 kDa. The protein migrates as 64 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8174 Supplier ACROBiosystems Supplier No. LA1C52A3100UG
Recombinant Protein;Rhesus macaque LAIR1 / CD305 Protein, Mouse IgG2a Fc Tag;100UG;Rhesus macaque LAIR-1, mouse IgG2a Fc tag (LA1-C52A3) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Try 165 (Accession # ALV66454.1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 42.9 kDa. The protein migrates as 64 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.