Learn More
Aviva Systems Biology Corp LAIR2 Antibody - N-terminal region (ARP42032_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42032_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LAIR2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LAIR2. Alternate names: CD306, MGC71634ApplicationWB. Species reactivity: Human
Catalog No. 50-014-58087