Learn More
ACROBiosystems Recombinant Protein;Human LAIR2 / CD306 Protein;HEK293;1MG;LA2,LAIR2,CD306,LAIR-2
Catalog No. 501658356
Encompass_Preferred
Recombinant Protein;Human LAIR2 / CD306 Protein;1MG;Human LAIR-2, His Tag (LA2-H5220) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Pro 152 (Accession # AAH69366).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.9 kDa. The protein migrates as 19-23 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8356 Supplier ACROBiosystems Supplier No. LA2H52201MG
Recombinant Protein;Human LAIR2 / CD306 Protein;1MG;Human LAIR-2, His Tag (LA2-H5220) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Pro 152 (Accession # AAH69366).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.9 kDa. The protein migrates as 19-23 kDa under reducing (R) condition (SDS-PAGE).