Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human LAIR2 / CD306 Protein, His Tag & Avi Tag (Avitag™);HEK293;200UG;LA2,LAIR2,CD306,LAIR-2
Supplier:  ACROBiosystems LA2H82E4200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human LAIR2 / CD306 Protein, His Tag & Avi Tag (Avitag™);200UG;Biotinylated Human LAIR-2, His Tag, Avi Tag (LA2-H82E4) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Pro 152 (Accession # Q6ISS4-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 17.8 kDa. The protein migrates as 25-28 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8278