Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lake Red CBA, TCI America™
SDP

Catalog No. L007425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L007425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L007425G Supplier TCI America Supplier No. L007425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5160-02-1
Molecular Formula C34H24BaCl2N4O8S2
Molecular Weight (g/mol) 888.93
MDL Number MFCD01941571
InChI Key JNIGYQOBELCEIZ-MAKDUZDQSA-L
Synonym 1-(4-Chloro-o-sulfo-5-tolylazo)-2-naphthol Barium Salt
PubChem CID 131842502
IUPAC Name barium;5-chloro-4-methyl-2-[(2Z)-2-(2-oxonaphthalen-1-ylidene)hydrazinyl]benzenesulfonic acid
SMILES CC1=CC(=C(C=C1Cl)S(=O)(=O)O)NN=C2C(=O)C=CC3=CC=CC=C32.CC1=CC(=C(C=C1Cl)S(=O)(=O)O)NN=C2C(=O)C=CC3=CC=CC=C32.[Ba]

Specifications

Quantity 25 g
UN Number 1564
Formula Weight 888.93
Physical Form Crystalline Powder
Chemical Name or Material Lake Red CBA
EINECSNumber (5)-3242
RTECSNumber DB5500000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.