Learn More
MSC Lakeland ChemMax 2 Coverall, Taped Seams, Elastic Wrists/Ankles, 1 ea
Catalog No. 5000687556
Encompass
ChemMax 2 is a mid-range chemical protective suit constructed with a Saranex 3P barrier film laminated to a bicomponent spunbonded nonwoven substrate. This design provides protection against various chemicals, suitable for applications like chemical mixing, handling, and hazardous materials remediation. It features a light, soft, and flexible fabric.
  • Saranex 3P barrier film for broad chemical protection
  • Stitched and taped seams ensure strength and impermeability
  • Double zip with storm flap for secure front fastening
  • Cushioned knee-pads for enhanced durability and comfort
  • Super-B style pattern offers ergonomic design and freedom of movement
  • Light, soft, and flexible fabric ensures wearer comfort
  • Certified to EN 13034 (Type 6), EN 13982 (Type 5), EN 14605 (Type 3 & 4), EN 1073, EN 14126, and EN 1149-5
Catalog No. 50-006-87556 Supplier MSC Supplier No. 53919791
ChemMax 2 is a mid-range chemical protective suit constructed with a Saranex 3P barrier film laminated to a bicomponent spunbonded nonwoven substrate. This design provides protection against various chemicals, suitable for applications like chemical mixing, handling, and hazardous materials remediation. It features a light, soft, and flexible fabric.
  • Saranex 3P barrier film for broad chemical protection
  • Stitched and taped seams ensure strength and impermeability
  • Double zip with storm flap for secure front fastening
  • Cushioned knee-pads for enhanced durability and comfort
  • Super-B style pattern offers ergonomic design and freedom of movement
  • Light, soft, and flexible fabric ensures wearer comfort
  • Certified to EN 13034 (Type 6), EN 13982 (Type 5), EN 14605 (Type 3 & 4), EN 1073, EN 14126, and EN 1149-5