Learn More
MSC Lakeland Coverall, Pyrolon Plus 2, Hooded, Elastic Cuff, 25/Pk
Catalog No. 5000673075
Encompass
Lakeland Pyrolon Plus 2 coveralls are breathable garments designed to repel water, grease, oil, and other hazards, particularly where flash fire is a concern. These secondary FR coveralls are intended to protect primary FR clothing from both hazardous and non-hazardous contaminants.
  • Attached hood
  • Zipper closure
  • Elastic wrists
  • Elastic ankles
  • Wet or dry strength superior to traditional flame resistant disposables
  • Breathable fabric for comfort
  • Suitable for hazardous or non-hazardous contaminants
  • Meets NFPA 2113 2020 edition requirements (section 5.1.10) and ANSI/ISEA 203 Certified
Catalog No. 50-006-73075 Supplier MSC Supplier No. 30874499
Lakeland Pyrolon Plus 2 coveralls are breathable garments designed to repel water, grease, oil, and other hazards, particularly where flash fire is a concern. These secondary FR coveralls are intended to protect primary FR clothing from both hazardous and non-hazardous contaminants.
  • Attached hood
  • Zipper closure
  • Elastic wrists
  • Elastic ankles
  • Wet or dry strength superior to traditional flame resistant disposables
  • Breathable fabric for comfort
  • Suitable for hazardous or non-hazardous contaminants
  • Meets NFPA 2113 2020 edition requirements (section 5.1.10) and ANSI/ISEA 203 Certified