Learn More
MSC Lakeland Industries Coverall, MicroMax NS, Cool Suit, Collared, 25/Pk
Catalog No. 5000801995
Encompass
MicroMax NS Cool Suit Coveralls are designed with a breathable back panel to allow hot air to escape, ensuring wearer comfort. Constructed from a high-quality microporous film laminate, they offer superior resistance against liquids, light oils, and light chemical sprays. These coveralls are ideal for general maintenance and protection against dirt, dust, grease, and grime.
  • Collar
  • Open wrists and ankles
  • Breathable back panel
  • High moisture vapor transmission rate for comfort
  • Serged seams
  • Ergonomic styling for optimized fit and freedom of movement
Catalog No. 50-008-01995 Supplier MSC Supplier No. 37207792
MicroMax NS Cool Suit Coveralls are designed with a breathable back panel to allow hot air to escape, ensuring wearer comfort. Constructed from a high-quality microporous film laminate, they offer superior resistance against liquids, light oils, and light chemical sprays. These coveralls are ideal for general maintenance and protection against dirt, dust, grease, and grime.
  • Collar
  • Open wrists and ankles
  • Breathable back panel
  • High moisture vapor transmission rate for comfort
  • Serged seams
  • Ergonomic styling for optimized fit and freedom of movement