Learn More
Lakeland Industries Pyrolon Plus 2 Coveralls
Catalog No. 190882269B
Click to view available options
Description:
Attached hood and boots
Size:
Medium
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Size
19-088-2269B Attached hood and boots Medium
19-088-2269D Attached hood and boots X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-088-2269B Supplier Lakeland Industries Supplier No. 7414BMD
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Size Medium
Material Polyester Wood Pulp Spunlace
Color Blue
Disposable Single-use
Ankle Style Attached Boots
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper
Seam Type Serged
Certifications/Compliance NFPA 2113, ANSI ISEA 101
Description Attached hood and boots
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov