Learn More
Lakeland Industries Pyrolon Plus 2 Coveralls
Catalog No. 190882269D
Click to view available options
Description:
Attached hood and boots
Size:
Medium
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Size
19-088-2269D Attached hood and boots X-Large
19-088-2269B Attached hood and boots Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-088-2269D Supplier Lakeland Industries Supplier No. 7414BXL
This item is not returnable. View return policy
Specifications
Size X-Large
Material Polyester Wood Pulp Spunlace
Color Blue
Disposable Single-use
Ankle Style Attached Boots
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper
Seam Type Serged
Certifications/Compliance NFPA 2113, ANSI ISEA 101
Description Attached hood and boots
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov