Learn More
Lakeland Industries Pyrolon Plus 2 Coveralls with Attached Hood
Catalog No. 19059860
Click to view available options
Size:
4X-Large
5X-Large
6X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-059-860 4X-Large
19-059-861 5X-Large
19-059-862 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-059-860 Supplier Lakeland Industries Supplier No. 7428B4X
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Size 4X-Large
Material Fabric
Color Blue
Disposable Single-use
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper
Seam Type Serged
Certifications/Compliance NFPA 2113, 201, ANSI ISEA 101
Description Breathable, Attached hood, zipper, elastic wrists, ankles
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov