Learn More
Lakeland Industries Pyrolon Plus 2 Coveralls with Collar
Catalog No. 19082090
Click to view available options
Size:
Large
Color:
Blue
Description:
With collar and zipper
Catalog No. Size Color Description
19-082-090 Large Blue With collar and zipper
1 options
This item is not returnable. View return policy
Catalog No. 19-082-090 Supplier Lakeland Industries Supplier No. 7412BLG
This item is not returnable. View return policy
Specifications
Size Large
Material Polyester Wood Pulp Spunlace
Color Blue
Disposable Single-use
Neck Style Collared
Closure Type Zipper
Seam Type Serged
Certifications/Compliance NFPA 2113, ANSI ISEA 101
Description With collar and zipper