Learn More
MSC Lakeland Pyrolon CRFR Coverall, Hooded, Elastic Cuffs, Secondary FR, 6/Case
Catalog No. 5000781901
Encompass
Lakeland Pyrolon CRFR coveralls are designed for situations requiring protection from moderate chemical exposure and flash fire. These secondary FR coveralls are commonly used in paint manufacturing, environmental clean-up, or oil and gas, protecting primary FR against flammable contaminants and dry particulates.
  • Attached hood
  • Elastic wrists and ankles
  • Storm flap over zipper
  • Supplied in cases of 6
Catalog No. 50-007-81901 Supplier MSC Supplier No. 48384127
Lakeland Pyrolon CRFR coveralls are designed for situations requiring protection from moderate chemical exposure and flash fire. These secondary FR coveralls are commonly used in paint manufacturing, environmental clean-up, or oil and gas, protecting primary FR against flammable contaminants and dry particulates.
  • Attached hood
  • Elastic wrists and ankles
  • Storm flap over zipper
  • Supplied in cases of 6