Learn More
MSC Lakeland Pyrolon Plus 2 FR Coverall, Collared, Open Wrist/Ankle, 25/Cs
Catalog No. 5000787367
Encompass
Lakeland Pyrolon Plus 2 coveralls are breathable garments designed to repel water, grease, oil, and other hazards, specifically for environments where flash fire is a concern. This secondary FR collared coverall features open wrists and ankles, intended to protect primary FR clothing from both hazardous and non-hazardous substances.
  • Zipper closure
  • Open wrists and ankles
  • Breathable garment
  • Repels water, grease, oil, and other hazards
  • Designed for flash fire concerns
  • Secondary FR collared coverall
Catalog No. 50-007-87367 Supplier MSC Supplier No. 86109345
Lakeland Pyrolon Plus 2 coveralls are breathable garments designed to repel water, grease, oil, and other hazards, specifically for environments where flash fire is a concern. This secondary FR collared coverall features open wrists and ankles, intended to protect primary FR clothing from both hazardous and non-hazardous substances.
  • Zipper closure
  • Open wrists and ankles
  • Breathable garment
  • Repels water, grease, oil, and other hazards
  • Designed for flash fire concerns
  • Secondary FR collared coverall