Learn More
Hycult Biotech Inc LAL CHROMOGENIC ENDPOINT ASSAY
SDP
Catalog No. NC0341975
Encompass
The LAL assays is to be used for the in vitro quantitative determination of bacterial endotoxin in various biological fluids (including sera), devices, (air)filters and tissue culture medium.
Catalog No. NC0341975 Supplier Hycult Biotech Inc Supplier No. HIT302
The LAL assays is to be used for the in vitro quantitative determination of bacterial endotoxin in various biological fluids (including sera), devices, (air)filters and tissue culture medium.