Learn More
FUJIFILM Biosciences LAL REAGENT WATER 12X100ML
Catalog No. 5000450413
Encompass
endotoxin-free water with 0 001EU/mL of endotoxin for use in reconstitution and dilution
Catalog No. 50-004-50413 Supplier FUJIFILM Biosciences Supplier No. LRW12100
endotoxin-free water with 0 001EU/mL of endotoxin for use in reconstitution and dilution