Learn More
Charles River Laboratories LAL Reagent Water; 30mL bottle, 12/case
Catalog No. NC9490719
Encompass
LAL Reagent Water; 30mL bottle; (<0.001 EU/mL) 12/case
Catalog No. NC9490719 Supplier Charles River Laboratories Supplier No. W130
LAL Reagent Water; 30mL bottle; (<0.001 EU/mL) 12/case