Learn More
Associates of Cape Cod Inc LAL Reagent Water (LRW), 500 mL/Plastic bottle, 12 bottles/pack
SDP
Catalog No. NC0751166
Encompass
LRW is specially processed water intended for reconstitution of LAL reagents and CSE, and to dilute samples and standards for LAL assays. Not for human or animal injection. LRW contains less than 0.001 EU/mL endotoxin and less than 1.56 pg/mL glucan. Plastic vial w/ screw cap closure, 500 mL/vial, 12 vials per case
Catalog No. NC0751166 Supplier Associates of Cape Cod Inc Supplier No. WP500C
LRW is specially processed water intended for reconstitution of LAL reagents and CSE, and to dilute samples and standards for LAL assays. Not for human or animal injection. LRW contains less than 0.001 EU/mL endotoxin and less than 1.56 pg/mL glucan. Plastic vial w/ screw cap closure, 500 mL/vial, 12 vials per case