Learn More
Aviva Systems Biology Corp LAMA4 Antibody - C-terminal region (ARP54550_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54550_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LAMA4. It was validated on Western Blot. . Alternate names: DKFZp686D23145, LAMA3, LAMA4*-1ApplicationWB. Species reactivity: Human
Catalog No. 50-285-937