Learn More
Aviva Systems Biology Corp LAMB3 Antibody - middle region (ARP41694_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41694_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LAMB3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LAMB3. Alternate names: LAMNB1, LAM5, BM600-125KDAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Rabbit
Catalog No. 50-014-57954