Learn More
Bioworld Lambda Broth, 500 g
Catalog No. 50254428
Encompass_Preferred
Lambda BrothMedia for the propagation of lambda phageBiotechnology Grade pH: 6.9Dehydrated Appearance: light beige powder Soluble in water
Catalog No. 50-254-428 Supplier Bioworld Supplier No. 306210052
Lambda BrothMedia for the propagation of lambda phageBiotechnology Grade pH: 6.9Dehydrated Appearance: light beige powder Soluble in water