Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

lambda-Carrageenan (High-viscosity), TCI America™
SDP

Catalog No. C331325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C331325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C331325G Supplier TCI America Supplier No. C331325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 9064-57-7
Molecular Formula C12H19O20S3-3
Molecular Weight (g/mol) 579.444
MDL Number MFCD00151513
InChI Key UWPXLSAITSWCRB-UHFFFAOYSA-K
PubChem CID 91972149
IUPAC Name [5-[4,5-dihydroxy-6-(hydroxymethyl)-3-sulfonatooxyoxan-2-yl]oxy-2,4-dihydroxy-6-(sulfonatooxymethyl)oxan-3-yl] sulfate
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)OS(=O)(=O)[O-])O)COS(=O)(=O)[O-])OS(=O)(=O)[O-])O)O)O

Specifications

Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material lambda-Carrageenan (High-viscosity)
RTECSNumber FI0704000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.