Learn More
New England Biolabs, Inc. Lambda DNA – 250 µg
SDP
Catalog No. 50811747
Encompass
Lambda DNA is a commonly used DNA substrate. It is duplex DNA is isolated from bacteriophage lambda (cI857ind 1 Sam 7). Lambda DNA is 48,502 base pairs in length.
Catalog No. 50-811-747 Supplier New England Biolabs, Inc. Supplier No. N3011S
Lambda DNA is a commonly used DNA substrate. It is duplex DNA is isolated from bacteriophage lambda (cI857ind 1 Sam 7). Lambda DNA is 48,502 base pairs in length.