Learn More
New England Biolabs, Inc. Lambda DNA-Hind III Digest – 0.3 ml
SDP
Catalog No. 50811749
Encompass
The HindIII digest of lambda DNA (cI857ind1 Sam 7) yields 8 fragments suitable for use as molecular weight standards for agarose gel electrophoresis. The approximate mass of DNA in each of the bands is provided (assuming a 1.0 ug load) for approximating the mass of DNA in comparably intense samples of similar size. Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.
Catalog No. 50-811-749 Supplier New England Biolabs, Inc. Supplier No. N3012S
The HindIII digest of lambda DNA (cI857ind1 Sam 7) yields 8 fragments suitable for use as molecular weight standards for agarose gel electrophoresis. The approximate mass of DNA in each of the bands is provided (assuming a 1.0 ug load) for approximating the mass of DNA in comparably intense samples of similar size. Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.