Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Lambda™ EliteMax Semi-automated Benchtop Pipettor

Catalog No. 07202490 Shop All Corning Products
Catalog No. 07-202-490 Supplier Corning™ Supplier No. 6070

missing translation for 'validMainframeMsg'

Plate filling from reagent reservoirs

The Corning™ Lambda™ EliteMax Semi-automated Benchtop Pipettor can perform a variety of liquid handling tasks including:

  • Plate filling from reagent reservoirs
  • Serial dilutions, plate-to-plate transfers, tube-to-tube transfers, and plate replication
  • Well-to-well transfers (cherry picking protocol for single-channel head only). Unit comes with a single-channel and 12-channel head installed.
  • Accurately and precisely transferring liquids and compounds between microplates
  • Transferring by row or by column

More complex applications can be performed such as sample preparation, timed reaction, PCR preparation, and cell-based assays set up.

TRUSTED_SUSTAINABILITY

Specifications

Accuracy 20 μL < ±2.0%; 100 μL < ±1.5%; 200 μL < ±1.0%
Product Type Semi-Automated Benchtop Pipettor
Device Holds Single-channel, 12-channel, 24-channel and custom-channel heads
Volume (Metric) 1 to 200 μL
For Use With (Equipment) Corning™ Lambda™ EliteMax pipet tips
Includes Corning™ Lambda™ EliteMax Semi-automated Benchtop Pipettor, Touch screen controller with software and license, External power supply, Portrait adapter stainless steel plate, Power cords (US, EU, UK, CH), Instruction manual, Calibration certificate
Plate Type 96-well standard, shallow, deep well, microcentrifuge, V-, U-, and flat-bottom
Precision 20 μL <1.5% CV; 100 μL <1.0% CV; 200 μL <0.7% CV
Temperature (Metric) 5°C to 40°C (relative humidity 80°C% up to 30°C or 45% relative humidity at 40°C)
Voltage 100/240 V
Frequency 47 to 63 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.