Learn More
Corning™ Lambda™ EliteTouch™ Pipettor Starter Kit
GREENER_CHOICE
Catalog No. 07201055 Shop All Corning Products
Change view
Click to view available options
Number of Channels:
1
Catalog No. Number of Channels
07-201-055 1
1 options
This item is not returnable. View return policy
Catalog No. 07-201-055 Supplier Corning™ Supplier No. 6065
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Corning™ Lambda™ EliteTouch™ Pipettor Starter Kit has been engineered to provide the highest levels of ergonomics, accuracy, and precision.
Corning Lambda EliteTouch Starter Kit contains 4 variable volume pipettors in the most popular and universal volumes (0.5-10 μL, 2-20 μL, 20-200 μL, and 100-1000 μL), pipet stand for 4 pipettors, 3 racks with tips, and accessories.
  • Cat. No. 6051 Lambda™ EliteTouch™ Single-channel pipettor, 0.5 to 10 μL
  • Cat. No. 6052 Lambda EliteTouch Single-channel pipettor, 2 to 20 μL
  • Cat. No. 6055 Lambda EliteTouch Single-channel pipettor, 20 to 200 μL
  • Cat. No. 6056 Lambda EliteTouch Single-channel pipettor, 100 to 1000 μL
  • Cat. No. 6066 Universal linear stand for four single-channel pipettors
  • Cat. No.4134 (10 μL) Corning DeckWorks pipet tips
  • Cat. No. 4138 (200 μL) Corning DeckWorks pipet tips
  • Cat. No. 4140 (1000 μL) Corning DeckWorks pipet tips
  • 3 colored pushbuttons (4 sets)
Learn more from North America
Learn more from Europe, Middle East, and Africa
See resource tab below for specifications.
'
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO8655
Autoclavable Autoclavable
Grip Type Ergonomic
Number of Channels 1
Product Line Lambda EliteTouch
Type Starter Kit
Finger Style Ergonomic
Includes 4 single-channel pipettors, universal linear stand for four single-channel pipettors, DeckWorks pipet tips, 3 colored pushbuttons (4 sets), accessories
Quantity 1
Warranty 3 Years
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov