Learn More
Apexbio Technology LLC Lambda Exonuclease(Synonyms: Lambda Exo, Lambda exonuclease enzyme, λ exonuclease, λ exonuclease enzyme), 1000U.
SDP
Supplier:  Apexbio Technology LLC K10971000
Encompass_Preferred
Lambda Exonuclease is a DNA-specific exonuclease enzyme that progressively degrades double-stranded DNA in a 5 3 direction preferentially initiating digestion at 5 -phosphorylated termini It displays minor activity toward single-stranded DNA and double-stranded DNA with non-phosphorylated ends but cannot start cleavage at internal nicks or gaps Lambda Exonuclease is commonly employed in molecular biology laboratories for the generation of single-stranded DNA templates DNA single-strand conformation polymorphism (SSCP) analysis selective degradation of linear plasmid forms while preserving supercoiled structures and rolling-circle amplification protocols
Catalog No. 50-000-61387