Learn More
MOLECULAR DEPOT LLC Lambda FLC Antibody - 1 mg
Catalog No. 5000013523
Encompass_Preferred
Lambda Free Light Chains Antibody (1 mg) consists of affinity-purified polyclonal antibodies specific to human Lambda Free Light Chains Tested in several immunoassay platforms including ELISA CLIA Immunoturbidimetry and more Readily available in bulk amounts and at bulk prices Biotinylation and other labeling services are also available For in vitro research use only
Catalog No. 50-000-13523 Supplier MOLECULAR DEPOT LLC Supplier No. A2010037
Lambda Free Light Chains Antibody (1 mg) consists of affinity-purified polyclonal antibodies specific to human Lambda Free Light Chains Tested in several immunoassay platforms including ELISA CLIA Immunoturbidimetry and more Readily available in bulk amounts and at bulk prices Biotinylation and other labeling services are also available For in vitro research use only