Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lambda Light Chain Antibody (RM110), Novus Biologicals™
SDP

Catalog No. NBP261910
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61910 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61910 Supplier R&D Systems Supplier No. NBP261910

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Monoclonal Antibody

Lambda Light Chain Monoclonal specifically detects Lambda Light Chain in Mouse samples. It is validated for ELISA, Western Blot.

Specifications

Antigen Lambda Light Chain
Applications ELISA, Western Blot
Classification Monoclonal
Clone RM110
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 0.1 - 0.5 ug/mL, Flow Cytometry, ELISA 0.005 ug/ml - 0.2 ug/ml, Immunohistochemistry 0.5 - 2 ug/mL, Immunoprecipitation, Immunocytochemistry 0.5 - 2 ug/mL
Formulation PBS, 50% Glycerol, 1% BSA with 0.09% Sodium Azide
Gene Alias IGLC, immunoglobulin lambda constant group
Gene Symbols IGLC1
Host Species Rabbit
Immunogen Mouse IgM lambda
Purification Method Protein A purified
Quantity 100 μg
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3536
Test Specificity This antibody reacts to the lambda light chain of mouse immunoglobulins. No cross reactivity with the kappa light chain, human IgG, rat IgG, or goat IgG.
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.