Learn More
Diagnostic Biosystems LAMBDA LIGHT CHAIN-FITC Lambda Light Chain-FITC
Supplier:  Diagnostic Biosystems F002
Encompass_Preferred
Traces of contaminating antibodies have been removed by solid phase absorption. Only the lambda precipitation line appeared in cross immunoelectrophoresis. This antibody is well suited for tissue section staining but may also be used for other applications.
Catalog No. 50-240-7379