Learn More
Sigma Aldrich Fine Chemicals Biosciences Lambda Light Chains free f1MG
Catalog No. 502904790
Encompass
Immunoglobulins (Ig) are produced by the B cells and play crucial role in host humoral immune response. A typical immunoglobulin consists of two heavy chains and two light chains. The two light chains are eitherº or the» chain. The B cells express one type of light chain during their life time» Light Chains (free) from human myeloma sera consists of IgG» light chains purified from myeloma sera.
Catalog No. 50-290-4790 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L06651MG
Immunoglobulins (Ig) are produced by the B cells and play crucial role in host humoral immune response. A typical immunoglobulin consists of two heavy chains and two light chains. The two light chains are eitherº or the» chain. The B cells express one type of light chain during their life time» Light Chains (free) from human myeloma sera consists of IgG» light chains purified from myeloma sera.